missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Power Cords for myECL Imager and Pierce G2 Fast Blotter

Product Code. 13403706 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
C/13 Power Cord, EU
C/13 Power Cord, UK
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
13403706 C/13 Power Cord, UK Each
13395081 C/13 Power Cord, EU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13403706 Supplier Thermo Scientific™ Supplier No. 84860

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These electrical power cords have a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets of specific countries.

Thermo Scientific™ Power Cords for myECL Imager and Pierce G2 Fast Blotter have a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets of specific countries.

Additional Information and Online Ordering

The black, 2.4- to 3.5m-long, cords have been validated to replace the cordset that is supplied with particular Thermo Scientific powered devices. Specifically, these instruments include the myECL Imager (Part No. 62236) and the Pierce G2 Fast Blotter (Part No. 62288).

Highlights:

  • Black
  • 2.4 to 3.5m long
  • C13 connector
  • Country-specific connectors
TRUSTED_SUSTAINABILITY

Specifications

Product Type C/13 Power Cord, UK
Length (English) 94.48 to 137 in.
Length (Metric) 2.4 to 3.5 m
Formulation Black 2.5 m power cord
Color Black
For Use With (Application) Protein biology, protein gel electrophoresis, protein assays and analysis, western blotting
Sufficient For Replacement of myECL Imager and Pierce G2 Fast Blotter accessory
Quantity 1 Cord
For Use With (Equipment) Pierce Power System, myECL Imager
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.