missing translation for 'onlineSavingsMsg'
Learn More

Replacement Power Adapter 12V for Fisherbrand™ Mini Centrifuge

Product Code. 15895181 Shop All Heathrow Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand Mini-centrifuge
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15895181 Fisherbrand Mini-centrifuge Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15895181 Supplier Heathrow Scientific Supplier No. HS120491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Power adapter, for use with Fisherbrand™ Mini Centrifuge (16617645).

  • Color: Black

Specifications

Type Power Adapter
For Use With (Equipment) Fisherbrand Mini-centrifuge
Color Black
Certifications/Compliance UL, GS, TUV, CE, PSE, and C-Tick
Voltage 12 VDC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.