missing translation for 'onlineSavingsMsg'
Learn More

Cleaver Scientific Power Supply Adapters

Product Code. 16329085 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
No. per Pack:
2/Pk.
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
16329085 2/Pk. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16329085
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Connecting Cleaver Scientific Electrophoresis Tanks to 2 mm Input Power Supplies

  • Use these adapters for power supplies with 2 mm inputs
  • Cleaver Scientific systems typically have 4 mm connectors
  • Gold-plated connections

Specifications

For Use With (Application) Connecting Cleaver Scientific Electrophoresis Tanks to 2 mm Input Power Supplies
Type Gold Plated Connections
Model CSL-4-2
No. per Pack 2/Pk.
Size Input: 4 mm, Output: 2 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.