missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Power Supply Cable for Lynx Superspeed
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Centrifuge
Unit Size:
Each
Description
- IEC60309 16A-6h 5 pin Red (3P+N+PE), 380/400 V
Specifications
Specifications
| Type | Power Cable |
| For Use With (Equipment) | Centrifuge |
| Disposable | Reusable |
| Color | Red |
| Description | IEC60309 16A-6h 5 pin Red (3P+N+PE), 380/400 V |
| Voltage | 380/400 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction