Learn More
Fisherbrand™ Power Supply for Fisherbrand™ accumet™ AB and XL Benchtop Meters
Product Code. 12850653 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 Meters
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12850653 AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 Meters Each
1 options
Product Code. 12850653
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Universal replacement power supply
  • For use with accumet benchtop meters models: AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 meters
TRUSTED_SUSTAINABILITY
Specifications
Type Power Supply
Electrical Requirements 110/230 VAC, 9 VDC, 18 W, center positive
For Use With (Equipment) AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 Meters