missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PowerEase™ Touch 120W Power Supply and Mini Gel Tank with Mini Blot Module Set Welcome Pack

Product Code. 17144579
Change view
Click to view available options
Quantity:
1 pack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17144579 1 pack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17144579 Supplier Invitrogen™ Supplier No. PSC120MB
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PowerEase Touch 120W Power Supply and Mini Gel Tank with Mini Blot Module Set Welcome Pack contains: PowerEase Touch 120W Power Supply and Mini Gel Tank and Blot Module Set

The PowerEase Touch 120W Power Supply and Mini Gel Tank with Mini Blot Module Set Welcome Pack contains:

  • PowerEase Touch 120W Power Supply
  • Mini Gel Tank and Blot Module Set

About the PowerEase Touch 120W Power Supply

The Invitrogen PowerEase Touch 120W Power Supply is a medium-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for DNA/RNA electrophoresis, SDS-PAGE, and native PAGE. The PowerEase Touch 120W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the Mini Gel Tank and Blot Module Set

The Mini Gel Tank and Blot Module Set is an electrophoresis and blotting system for mini gels. The Mini Gel Tank is a vertical mini-gel electrophoresis system with a unique tank design that has convenient side-by-side gel loading and enhanced viewing during use. The tank can accommodate up to two gels per run with each gel run in its own separate chamber, making it easy to run just one gel. The tank is compatible with all Invitrogen precast mini gels and SureCast handcast gels.

The Mini Blot Module is a wet transfer device used exclusively with the Mini Gel Tank. The tank accommodates one blot module per chamber, or two blot modules total with the side-by-side layout. The universal connection and molded gasket make the blot module easy to use, while the inner core of the blot module allows for use of less methanol-based transfer buffer per wet transfer. At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes typically in 30 to 60 minutes.

TRUSTED_SUSTAINABILITY

Specifications

Ambient Temperature (English) Room Temperature
For Use With (Application) DNA/RNA electrophoresis, SDS-PAGE, and native PAGE
Gel Size Mini
Max. Current 500 mA
Product Line PowerEase
Timer Up to 999 min./step
Display 4.3 inch backlit LCD
No. of Jacks 4 (4 positive voltage and 4 negative voltage)
Quantity 1 pack
Shipping Condition Room temperature
Stackable Yes
Warranty One year
Dimensions (L x W x H) 30.3 inch x 23 inch x 10.2 inch
Weight 2.1 kg
Max. Voltage 300 VDC
Wattage 120 watts
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.