missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PowerEase™ Touch 350W Power Supply and and Mini Gel Tank with Mini Blot Module Set Welcome Pack

Product Code. 17159540
Change view
Click to view available options
Quantity:
1 pack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17159540 1 pack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17159540 Supplier Invitrogen™ Supplier No. PSC350MB
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PowerEase Touch 350W Power Supply and Mini Gel Tank with Mini Blot Module Set Welcome Pack contains: PowerEase Touch 350W Power Supply and Mini Gel Tank and Blot Module Set

The PowerEase Touch 350W Power Supply* and Mini Gel Tank with Mini Blot Module Set Welcome Pack contains:

• PowerEase Touch 350W Power Supply, 115 VAC (Cat. No. PS0350)*
• PowerEase Touch 350W Power Supply, 230 VAC (Cat. No. PS0351)*
• PowerEase Touch 350W Power Supply, 230 VAC (Cat. No. PS0352)* (Australia)
• PowerEase Touch 350W Power Supply, 230 VAC (Cat. No. PS0352)* (China)
• Mini Gel Tank and Blot Module Set (Cat. No. NW2000)

*dependent on region

About the PowerEase Touch 350W Power Supply
The Invitrogen PowerEase Touch 350W Power Supply is a high-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for high current applications like high throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting. The PowerEase Touch 350W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the Mini Gel Tank and Blot Module Set
The Mini Gel Tank and Blot Module Set is an electrophoresis and blotting system for mini gels. The Mini Gel Tank is a vertical mini-gel electrophoresis system with a unique tank design that has convenient side-by-side gel loading and enhanced viewing during use. The tank can accommodate up to two gels per run with each gel run in its own separate chamber, making it easy to run just one gel. The tank is compatible with all Invitrogen precast mini gels and SureCast handcast gels.

The Mini Blot Module is a wet transfer device used exclusively with the Mini Gel Tank. The tank accommodates one blot module per chamber, or two blot modules total with the side-by-side layout. The universal connection and molded gasket make the blot module easy to use, while the inner core of the blot module allows for use of less methanol-based transfer buffer per wet transfer. At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes typically in 30 to 60 minutes.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Gel Electrophoresis
Gel Size Mini
Product Line PowerEase
Quantity 1 pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.