missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PowerEase™ Touch 350W Power Supply and SureLock™ Tandem Midi PVDF Welcome Pack

Product Code. 17185691
Change view
Click to view available options
Quantity:
1 pack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17185691 1 pack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17185691 Supplier Invitrogen™ Supplier No. PSC350STMPV
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PowerEase Touch 350W Power Supply and SureLock Tandem Midi PVDF Welcome Pack contains: PowerEase Touch 350W Power Supply, SureLock Tandem Midi Gel Tank, Two SureLock Tandem Midi Blot Modules, SureLock Tandem Transfer Tray and SureLock Tandem Midi Pre-cut Membranes and Filter, 0.45 μm, PVDF

The PowerEase Touch 350W Power Supply and SureLock Tandem Midi PVDF Welcome Pack contains:

  • PowerEase Touch 350W Power Supply
  • SureLock Tandem Midi Gel Tank
  • Two SureLock Tandem Midi Blot Modules
  • SureLock Tandem Transfer Tray
  • SureLock Tandem Midi Pre-cut Membranes and Filter, 0.45 μm, PVDF

About the PowerEase Touch 350W Power Supply

The Invitrogen PowerEase Touch 350W Power Supply is a high-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for high current applications like high throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting. The PowerEase Touch 350W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the SureLock Tandem Midi PVDF Welcome Pack

The SureLock Tandem Midi Welcome Pack, PVDF, is designed specifically for electrophoresis and wet transfer of Invitrogen pre-cast midi gels using PVDF transfer membranes. The innovative 2-in-1 gel electrophoresis and transfer tank enables electrophoresis and transfer of high-performance Invitrogen midi gels using the same tank.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Gel Electrophoresis
Gel Size Midi
Product Line PowerEase
Quantity 1 pack
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.