missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PowerEase™ Touch HV Power Supply Adapters

Product Code. 17882994
Change view
Click to view available options
Quantity:
1 set
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17882994 1 set Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17882994 Supplier Invitrogen™ Supplier No. PSHV00A
  • ONLINE EXCLUSIVE PRICE
    111.00 €
    89.91 € / Pack of 1
    Save 21.09 €
    19% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Invitrogen PowerEase Touch HV Power Supply Adapters are designed to facilitate the connection of some electrophoresis tanks to the PowerEase Touch HV Power Supply.

The Invitrogen PowerEase Touch HV Power Supply Adapters are designed to facilitate the connection of some electrophoresis tanks to the PowerEase Touch HV Power Supply. Four sets of adapters are included with each HV power supply, but additional sets are offered here for separate purchase.

Supported electrophoresis tanks include:
• ZOOM IPGRunner Mini-cell (Cat. No. ZM0001)
• XCell SureLock Mini-Cell (Cat. No. EI0001)
• Mini Gel Tank (Cat. No. A25977)
• XCell4 SureLock Midi-Cell (Cat. No. WR0100)

The adapters are designed to remain on the electrophoresis tank electrical leads when the lid is removed from the power supply.

The SureLock Tandem Midi Gel Tank (Cat. No. STM1001) does not require these adapters to be compatible with the PowerEase Touch HV power supply.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Gel Electrophoresis, Isoelectric Focusing (IEF or IPG)
For Use With (Equipment) XCell SureLock Mini, ZOOM IPGRunner Mini, PowerEase Touch HV Power Supply, XCell4 SureLock Midi, Mini Gel Tank
Product Line PowerEase
Quantity 1 set
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.