missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 80-100 Heavy Duty Crinkle Latex Coated Gloves

Product Code. 11766673
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11766673 9 12 pairs
11726673 10 12 pairs
11736673 11 12 pairs
11746673 7 12 pairs
11756673 8 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11766673 Supplier Ansell Supplier No. 80100090
  • ONLINE EXCLUSIVE PRICE
    74.66 €
    49.02 € / 12 pairs
    Save 25.64 €
    34% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Automatic knit construction delivers an optimally tensioned inner surface for superb fit and flexibility. Latex coating features a raised crinkle finish for enhanced grip in wet and dry conditions

  • Gloves deliver overall mechanical performance for maximal protection and durability
  • Latex coating's raised crinkle finish enhances grip in both dry and wet conditions
  • Automatic knitted liner delivers an optimally tensioned inner surface for improved fit and flexibility
  • Designed for use in a wide range of handling and carrying applications
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Crinkled Latex
Color Gray, Blue
Cut Resistance Rating ANSI A2
Exterior Finish Partially Textured
Hand Specific
Latex Free No
Material Polycotton
Product Type Dip-coated Gloves
Size 9
For Use With (Application) Machinery and Equipment
Glove Cuff Style Knit
Series 80-100
Yarn Gauge 10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.