missing translation for 'onlineSavingsMsg'
Learn More

Poziotinib, TRC

Product Code. 30419075
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419075 1 mg -
30410980 5 mg 5mg
30417463 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30419075 Supplier Toronto Research Chemicals Supplier No. TRCP2157701MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092364-38-9
Molecular Formula C23H21Cl2FN4O3
Molecular Weight (g/mol) 491.34
Synonym 1-[4-[[4-[(3,4-Dichloro-2-fluorophenyl)amino]-7-methoxy-6-quinazolinyl]oxy]-1-piperidinyl]-2-propen-1-one, 1-[4-[4-(3,4-Dichloro-2-fluoroanilino)-7-methoxyquinazolin-6-yl]oxypiperidin-1-yl]prop-2-en-1-one
IUPAC Name 1-[4-[4-(3,4-dichloro-2-fluoroanilino)-7-methoxyquinazolin-6-yl]oxypiperidin-1-yl]prop-2-en-1-one
SMILES COc1cc2ncnc(Nc3ccc(Cl)c(Cl)c3F)c2cc1OC4CCN(CC4)C(=O)C=C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C23H21Cl2FN4O3/c1-3-20(31)30-8-6-13(7-9-30)33-19-10-14-17(11-18(19)32-2)27-12-28-23(14)29-16-5-4-15(24)21(25)22(16)26/h3-5,10-13H,1,6-9H2,2H3,(H,27,28,29)
Formula Weight 490.0975
Chemical Name or Material Poziotinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.