missing translation for 'onlineSavingsMsg'
Learn More

Foxx Life Sciences PP Aspirator Bottle with Spigot

Product Code. 31933494
Encompass_Preferred
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31933494 20 L Each
31933364 5 L Each
31933437 10 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31933494 Supplier Foxx Life Sciences Supplier No. 155L897FLS
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for efficient liquid storage, handling and dispensing.

  • Features:
    • Manufactured from high-purity virgin USP Class VI Medical Grade polypropylene
    • Strong, durable construction for extended use
    • Leak-proof design with inner cap for a tight seal
    • High-quality, durable aspirator bottle with spigot/tap for controlled dispensing
    • Aspirator bottle for laboratory storage applications
    • Suitable for temperatures below 0°C
    • Autoclavable aspirator bottles

Specifications

Capacity (Metric) 20 L
Color Opaque White
Material Polypropylene
Sterility Non-sterile
Autoclavable Autoclavable
Product Type Aspirator Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.