missing translation for 'onlineSavingsMsg'
Learn More

Atago™ PR-100SA Model Digital Refractometer

Product Code. 11406288
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
11406288 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11406288 Supplier Atago™ Supplier No. 3488
  • ONLINE EXCLUSIVE PRICE
    765.00 €
    757.09 € / Pack of 1
    Save 7.91 €
    1% Off
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Type Digital Refractometer
Scale Range 0 to 100 Salinity
Model PR-100SA
Accuracy ±2‰
Resolution 1‰
Scale Type Concentration of salinity
For Use With (Application) Ideal for Salinity
IP Rating IP64
Weight (Metric) 300 g
Temperature Range (Metric) 5°C to 40°C
Power Supply Details 006P dry battery (9V)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.