missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pralidoxime chloride, MedChemExpress
Description
Pralidoxime chloride is a potent reactivator of acetylcholinesterase (AChE). Pralidoxime chloride reactivates nerve agent-inhibited AChE via direct nucleophilic attack by the oxime moiety on the phosphorus center of the bound nerve agent. Pralidoxime chloride is an antidote for organophosphate poisoning.
Specifications
Specifications
| Chemical Name or Material | Pralidoxime chloride |
| CAS | 51-15-0 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C7H9ClN2O |
| Quantity | 100 mg |
| Synonym | 2-PAM chloride |
| Solubility Information | DMSO : 21.67 mg/mL (125.54 mM; Need ultrasonic) |
| SMILES | C[N+]1=CC=CC=C1/C=N/O.[Cl-] |
| Molecular Weight (g/mol) | 172.61 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?