missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium foil, 1.0mm (0.04in) thick, 99.5% (REO)

Product Code. 11351479
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351479 25 x 25 mm 25mm
11490380 50 x 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351479 Supplier Thermo Scientific Alfa Aesar Supplier No. 013963.FF
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-10-0 | Pr | 140.91 g/mol

Praseodymium is used as an alloying constituent for alloys used to make permanent magnets and flints. Along with neodymium, praseodymium is used to manufacture yellow glass which can be used as eye protection (e.g. for welders).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.91
MDL Number MFCD00011174
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]

Spezifikation

Color White
Quantity 25 x 25 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈4.23g/25x25mm
Form Foil
Solubility Information Insoluble in water.
Percent Purity 99.5%
Assay (REO)
Chemical Name or Material Praseodymium foil, 1.0mm (0.04 in.) thick

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.