missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific™

Product Code. 11394118
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394118 50 g 50g
11384118 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394118 Supplier Thermo Scientific Alfa Aesar Supplier No. 041418.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For vacuum deposition

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-46-1
Molecular Formula F3Pr
Molecular Weight (g/mol) 197.90
MDL Number MFCD00016287
InChI Key BOTHRHRVFIZTGG-UHFFFAOYSA-K
IUPAC Name praseodymium(3+) trifluoride
SMILES [F-].[F-].[F-].[Pr+3]

Specifications

Melting Point 1395°C
Color Green
Physical Form Pieces
Quantity 50 g
UN Number UN3288
Solubility Information Soluble moderately in strong mineral acids Insoluble in water.
Formula Weight 197.9
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.2 g/mL
Assay (metals basis)
Chemical Name or Material Praseodymium(III) fluoride, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.