missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) nitrate hydrate, 99.9% (REO)

Product Code. 11397637
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Unit Size:
10g
1kg
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397637 50 g 50g
11317647 10 g 10g
11307647 250 g 250g
11387637 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11397637 Supplier Thermo Scientific Alfa Aesar Supplier No. 012909.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 15878-77-0 | H12N3O15Pr | 435.01 g/mol

Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15878-77-0
Molecular Formula H12N3O15Pr
Molecular Weight (g/mol) 435.01
MDL Number MFCD00011175
InChI Key LXXCECZPOWZKLC-UHFFFAOYSA-N
Synonym praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrate
PubChem CID 204170
SMILES O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Green
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99.9%
Solubility Information Soluble in water,anhydrous amine,alcohol,ether,polar solvents and acetonitrile.
IUPAC Name praseodymium(3+) hexahydrate trinitrate
Formula Weight 326.92 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Green crystals
Chemical Name or Material Praseodymium(III) nitrate hydrate, Rare Earth Oxides Content
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.