missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) Triflate, TRC

Product Code. 30424251
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424251 5 g 5g
30422054 2.5 g 2.5g
30418450 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424251 Supplier Toronto Research Chemicals Supplier No. TRCP7011935G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52093-27-3
Molecular Formula C3F9O9PrS3
Molecular Weight (g/mol) 588.11
Synonym Praseodymium Tris(trifluoromethanesulfonate), Praseodymium(3+) Tris(trifluoromethyl Sulfate), Praseodymium(III) Trifluoromethanesulfonate, Trifluoromethanesulfonic Acid Praseodymium(3+) Salt
IUPAC Name praseodymium-141(3+);trifluoromethanesulfonate
SMILES [141Pr+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 5 g
InChI Formula InChI=1S/3CHF3O3S.Pr/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3/i;;;1+0
Formula Weight 587.7637
Chemical Name or Material Praseodymium(III) Triflate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.