missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Product Code. 11313868
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313868 10 g 10g
11323868 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313868 Supplier Thermo Scientific Alfa Aesar Supplier No. 040310.09
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52093-27-3
Molecular Formula C3F9O9PrS3
Molecular Weight (g/mol) 588.097
MDL Number MFCD00192417
InChI Key ROUBZIWQWFQCHU-UHFFFAOYSA-K
Synonym praseodymium iii trifluoromethanesulfonate, praseodymium iii triflate, praseodymium trifluoromethanesulfonate, praseodymium 3+ tritriflate, praseodymium iii trifluoromethane-sulfonate, praseodymium iii tris trifluoromethanesulfonate, praseodymium 3+ ion tritrifluoromethanesulfonate
PubChem CID 9894708
IUPAC Name praseodymium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Pr+3]

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 588.11
Percent Purity 98%
Chemical Name or Material Praseodymium(III) trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.