missing translation for 'onlineSavingsMsg'
Learn More

Prasugrel Thiolactone (Mixture of Diastereomers), TRC

Product Code. 30012270
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012270 100 mg 100mg
30010602 500 mg 500mg
30010534 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012270 Supplier Toronto Research Chemicals Supplier No. TRCP701165100MG
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150322-38-6
Molecular Formula C18 H18 F N O2 S
Molecular Weight (g/mol) 331.4
Synonym Thieno[3,2-c]pyridin-2(4H)-one, 5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-5,6,7,7a-tetrahydro-, 5-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-5,6,7,7a-tetrahydrothieno[3,2-c]pyridin-2(4H)-one, 2-Oxo-5-(α-cyclopropylcarbonyl-2-fluorobenzyl)-2,4,5,6,7,7a-hexahydrothieno[3,2-c]pyridine, R 95913, Prasugrel Thiolactone, (7aRS)-5-[(1RS)-2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-5,6,7,7a-tetrahydrothieno[3,2-c]pyridin-2(4H)-one
IUPAC Name 5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4,6,7,7a-tetrahydrothieno[3,2-c]pyridin-2-one
SMILES Fc1ccccc1C(N2CCC3SC(=O)C=C3C2)C(=O)C4CC4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H18FNO2S/c19-14-4-2-1-3-13(14)17(18(22)11-5-6-11)20-8-7-15-12(10-20)9-16(21)23-15/h1-4,9,11,15,17H,5-8,10H2
Formula Weight 331.1042
Percent Purity >95%
Chemical Name or Material Prasugrel Thiolactone (mixture of diastereomers)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.