missing translation for 'onlineSavingsMsg'
Learn More

Prasugrel Thiolactone (Mixture of Diastereomers), TRC

Product Code. 30010602
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010602 500 mg 500mg
30012270 100 mg 100mg
30010534 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010602 Supplier Toronto Research Chemicals Supplier No. TRCP701165500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150322-38-6
Formula Weight 331.1042
InChI Formula InChI=1S/C18H18FNO2S/c19-14-4-2-1-3-13(14)17(18(22)11-5-6-11)20-8-7-15-12(10-20)9-16(21)23-15/h1-4,9,11,15,17H,5-8,10H2
IUPAC Name 5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4,6,7,7a-tetrahydrothieno[3,2-c]pyridin-2-one
Molecular Formula C18 H18 F N O2 S
Molecular Weight (g/mol) 331.4
Percent Purity >95%
SMILES Fc1ccccc1C(N2CCC3SC(=O)C=C3C2)C(=O)C4CC4
Synonym Thieno[3,2-c]pyridin-2(4H)-one, 5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-5,6,7,7a-tetrahydro-, 5-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-5,6,7,7a-tetrahydrothieno[3,2-c]pyridin-2(4H)-one, 2-Oxo-5-(α-cyclopropylcarbonyl-2-fluorobenzyl)-2,4,5,6,7,7a-hexahydrothieno[3,2-c]pyridine, R 95913, Prasugrel Thiolactone, (7aRS)-5-[(1RS)-2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-5,6,7,7a-tetrahydrothieno[3,2-c]pyridin-2(4H)-one
Chemical Name or Material Prasugrel Thiolactone (mixture of diastereomers)
Grade HPLC
Show More Show Less

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.