missing translation for 'onlineSavingsMsg'
Learn More

Pravastatin 1,1,3,3-Tetramethylbutylamine, TRC

Product Code. 30137617
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137617 25 mg 25mg
30136636 10 mg 10mg
17953555 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137617 Supplier Toronto Research Chemicals Supplier No. TRCP70202525MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151006-14-3
Molecular Formula C23 H36 O7 . C8 H19 N
Molecular Weight (g/mol) 553.77
Synonym Pravastatin 1,1,3,3-Tetramethylbutylamine, 1-Naphthaleneheptanoic acid, 1,2,6,7,8,8a-hexahydro-β,δ,6-trihydroxy-2-methyl-8-(2-methyl-1-oxobutoxy)-, [1S-[1α(βS*,δS*),2α,6α,8β(R*),8aα]]-, compd. with 2,4,4-trimethyl-2-pentanamine (1:1) (9CI), 2-Pentanamine, 2,4,4-trimethyl-, [1S-[1α(βS*,δS*),2α,6α,8β(R*),8aα]]-1,2,6,7,8,8a-hexahydro-β,δ,6-trihydroxy-2-methyl-8-(2-methyl-1-oxobutoxy)-1-naphthaleneheptanoate (9CI)
IUPAC Name (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic acid;2,4,4-trimethylpentan-2-amine
SMILES CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)O)[C@@H]12.CC(C)(C)CC(C)(C)N

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H36O7.C8H19N/c1-4-13(2)23(29)30-20-11-17(25)9-15-6-5-14(3)19(22(15)20)8-7-16(24)10-18(26)12-21(27)28;1-7(2,3)6-8(4,5)9/h5-6,9,13-14,16-20,22,24-26H,4,7-8,10-12H2,1-3H3,(H,27,28);6,9H2,1-5H3/t13-,14-,16+,17+,18+,19-,20-,22-;/m0./s1
Formula Weight 553.3979
Chemical Name or Material Pravastatin 1,1,3,3-tetramethylbutylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.