missing translation for 'onlineSavingsMsg'
Learn More

Pravastatin Lactone, TRC

Product Code. 30137167
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137167 50 mg 50mg
17943555 5 mg 5mg
17921645 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137167 Supplier Toronto Research Chemicals Supplier No. TRCP70119050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85956-22-5
Molecular Formula C23 H34 O6
Molecular Weight (g/mol) 406.51
Synonym Butanoic acid, 2-methyl-, (1S,3S,7S,8S,8aR)-1,2,3,7,8,8a-hexahydro-3-hydroxy-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (2S)-, Butanoic acid, 2-methyl-, 1,2,3,7,8,8a-hexahydro-3-hydroxy-7-methyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester, [1S-[1α(R*),3β,7β,8β(2S*,4S*),8aβ]]-, (+)-Pravastatin lactone, Pravastatin lactone, R 414, SQ 31369, (1S,3S,7S,8S,8aR)-3-Hydroxy-8-[2-[(2R,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethyl]-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate, Pravastatin Sodium Imp. D (EP)
IUPAC Name [(1S,3S,7S,8S,8aR)-3-hydroxy-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C23H34O6/c1-4-13(2)23(27)29-20-11-16(24)9-15-6-5-14(3)19(22(15)20)8-7-18-10-17(25)12-21(26)28-18/h5-6,9,13-14,16-20,22,24-25H,4,7-8,10-12H2,1-3H3/t13-,14-,16+,17+,18+,19-,20-,22-/m0/s1
Formula Weight 406.2355
Percent Purity >95%
Chemical Name or Material Pravastatin Lactone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.