missing translation for 'onlineSavingsMsg'
Learn More

Praziquantel, 98%

Product Code. 15457449
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457449 25 g 25g
15447449 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457449 Supplier Thermo Scientific Alfa Aesar Supplier No. J66188.14
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic. | CAS: 55268-74-1 | C19H24N2O2 | 312.413 g/mol

Praziquantel is a potent anthelmintic used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Praziquantel is a potent anthelmintic used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic.

Solubility
Freely soluble in ethanol or dichloromethane. Slightly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55268-74-1
Molecular Formula C19H24N2O2
Molecular Weight (g/mol) 312.413
MDL Number MFCD00058531
InChI Key FSVJFNAIGNNGKK-UHFFFAOYSA-N
Synonym praziquantel, droncit, biltricide, pyquiton, cesol, azinox, embay 8440, cutter tape tabs, prasiquantel, praziquantelum
PubChem CID 4891
IUPAC Name 2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one
SMILES C1CCC(CC1)C(=O)N2CC3C4=CC=CC=C4CCN3C(=O)C2

Specifications

Melting Point 136°C to 138°C
Boiling Point 1377°C
Flash Point >110°C (230°F)
Packaging Glass bottle
Quantity 25 g
Beilstein 761557
Solubility Information Freely soluble in ethanol or dichloromethane. Slightly soluble in water
Formula Weight 312.41
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Praziquantel
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.