missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Prazosin hydrochloride

Product Code. 30001107
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001107 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001107 Supplier R&D Systems™ Supplier No. 0623/100
  • ONLINE EXCLUSIVE PRICE
    181.09 €
    161.70 € / 100mg
    Save 19.39 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α1 and α2B antagonist; also MT3 antagonist

α1 and α2B-adrenoceptor antagonist. Also a potent antagonist at the melatonin MT3 receptor (Ki = 10.2 nM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 19237-84-4
Quantity 100 mg
Product Line Tocris
Molecular Formula C19H22ClN5O4
Formula Weight 419.87
Purity >99%
Molecular Weight (g/mol) 419.87
Chemical Name or Material Prazosin hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.