missing translation for 'onlineSavingsMsg'
Learn More

Prazosin hydrochloride

Product Code. 15484249
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484249 250 mg 250mg
15474249 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484249 Supplier Thermo Scientific Alfa Aesar Supplier No. J61712.MD
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An alpha adrenoceptor antagonist | CAS: 19237-84-4 | C19H22ClN5O4 | 419.87 g/mol

Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin

Solubility
Soluble in dimethyl sulfoxide, ethanol, water, methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19237-84-4
Molecular Formula C19H22ClN5O4
Molecular Weight (g/mol) 419.87
MDL Number MFCD00058177
InChI Key WFXFYZULCQKPIP-UHFFFAOYSA-N
Synonym prazosin hydrochloride, prazosin hcl, minipress, vasoflex, peripress, furazosin hydrochloride, deprazolin, hypovase, hypovasole, pratsiol
PubChem CID 68546
ChEBI CHEBI:8365
IUPAC Name hydrogen 2-[4-(furan-2-carbonyl)piperazin-1-yl]-6,7-dimethoxyquinazolin-4-amine chloride
SMILES [H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1=CC=CO1

Specifications

Melting Point 285°C
Color White
Quantity 250 mg
Beilstein 4303561
Merck Index 14,7717
Solubility Information Soluble in dimethyl sulfoxide,ethanol,water,methanol.
Formula Weight 419.86
Physical Form Powder
Chemical Name or Material Prazosin hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.