missing translation for 'onlineSavingsMsg'
Learn More

pre-Calcitriol PTAD Adduct (Mixture of Diastereomers), TRC

Product Code. 30136702
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136702 2.5 mg 2.5mg
30137180 10 mg 10mg
30004042 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136702 Supplier Toronto Research Chemicals Supplier No. TRCC1445202.5MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86307-44-0
Molecular Formula C35 H49 N3 O5
Molecular Weight (g/mol) 591.78
Synonym 1H,5H-Cyclopenta[f][1,2,4]triazolo[1,2-a]cinnoline-1,3(2H)-dione, 11-(3,5-dihydroxy-2-methyl-1-cyclohexen-1-yl)-4a,6,6a,7,8,9,9a,11-octahydro-7-(5-hydroxy-1,5-dimethylhexyl)-6a-methyl-2-phenyl-, [4aR-[4aα,6aα,7α(R*),9aβ,11α(3S*,5R*)]]- (9CI)
IUPAC Name 7-(3,5-dihydroxy-2-methylcyclohexen-1-yl)-13-(6-hydroxy-6-methylheptan-2-yl)-14-methyl-4-phenyl-2,4,6-triazatetracyclo[7.7.0.02,6.010,14]hexadec-8-ene-3,5-dione
SMILES C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2C3=C[C@H](N4N([C@@H]3CC[C@]12C)C(=O)N(C4=O)c5ccccc5)C6=C(C)[C@@H](O)C[C@H](O)C6

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C35H49N3O5/c1-21(10-9-16-34(3,4)43)27-13-14-28-26-20-30(25-18-24(39)19-31(40)22(25)2)38-33(42)36(23-11-7-6-8-12-23)32(41)37(38)29(26)15-17-35(27,28)5/h6-8,11-12,20-21,24,27-31,39-40,43H,9-10,13-19H2,1-5H3/t21-,24-,27-,28+,29-,30+,31+,35-/m1/s1
Formula Weight 591.3672
Chemical Name or Material pre-Calcitriol PTAD Adduct
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.