missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Pre-cut Blank Labels for BBP3X, S3XXX, and i3300 Printers

Product Code. 15991001 Shop All Brady Products
Change view
Click to view available options
Color:
White
Yellow
Height (Metric):
72 mm
75 mm
152 mm
Width (Metric):
28.6 mm
57 mm
101 mm
Vertical Repeat (Metric):
82.6 mm
158.8 mm
Unit Size:
Pack of 175
Pack of 300
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Height (Metric) Width (Metric) Vertical Repeat (Metric) Unit Size
15991001 White 72 mm 28.6 mm 82.6 mm Pack of 300
15981001 White 75 mm 57 mm 82.6 mm Pack of 300
15911011 Yellow 75 mm 57 mm 82.6 mm Pack of 300
15971001 White 152 mm 101 mm 158.8 mm Pack of 175
15901011 Yellow 152 mm 101 mm 158.8 mm Pack of 175
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15991001 Supplier Brady™ Supplier No. 120932
  • ONLINE EXCLUSIVE PRICE
    146.00 €
    138.78 € / Pack of 300
    Save 7.22 €
    4% Off
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The material of these labels is ideal for indoor and outdoor facility labels.

  • Ideal for 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels
  • Vinyl material

Specifications

Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Color White
Finish Gloss
Material Vinyl (B-7569)
Print Technology Thermal Transfer
Sterility Non-sterile
For Use With (Application) Facility Identification, Hazard Communication Identification, Electrical Labeling
For Use With (Equipment) BBP™30, BBP™31, BBP™33, BBP™35 Multicolor, BBP™37 Multicolor, BradyPrinter i3300, S3000, S3100, and i5300 Industrial Label Printers
Labels per Row 1
Quantity 300 Labels
Shape Rectangle
Temperature Range (Metric) -20°C to +75°C
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 82.6 mm
Width (Metric) 28.6 mm
Height (Metric) 72 mm
Alternate Part Number B30-242-7569-WT
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.