missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pre-miR™ miRNA Precursor Molecules—Negative Control #2

Product Code. 10391225
Change view
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10391225 5 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10391225 Supplier Invitrogen™ Supplier No. AM17111
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-miR™ miRNA Precursor Molecules—Negative Control #2

A synthetic miRNA mimic for miRNA functional analysis. 5 nmol is supplied.

  • Optimized design results in correct strand utilization by RISC
  • Purified and ready to transfect

    Invitrogen™ Pre-miR™ miRNA Precursor Molecules are small, chemically modified, double-stranded RNA molecules designed to mimic endogenous mature miRNAs. This ready-to-use miRNA mimic can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. They enable detailed study of miRNA biological effects via gain-of-function experiments. Pre-miR™ Negative Control #2 is a random sequence Pre-miR™ molecule that has been extensively tested in human cell lines and tissues and validated to produce no identifiable effects on known miRNA function. NOTE: Pre-miR™ miRNA Precursors are not hairpin constructs and should not be confused with pre-miRNAs.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.
    Format Tube
    Control Type Negative Control
    RNAi Type miRNA Mimic
    Species Human, Mouse, Rat
    For Use With (Application) miRNA & Non-Coding RNA Analysis
    Label or Dye Unconjugated
    Product Line Ambion, Pre-miR
    Product Type miRNA Mimic
    Quantity 5 nmol
    Shipping Condition Room Temperature
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.