missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Precise™ 8%, Tris-Glycine, 5.0 mm, 12-well

Product Code. 11824604 Shop All Thermo Scientific Products
Change view
Click to view available options
Wells:
12-well
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells unitSize
11824604 12-well Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11824604 Supplier Thermo Scientific™ Supplier No. 25265

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Separation Range: 45 to 205 kd; Separation Type: Denaturing, Native; For Use With: Novex XCell I and II; Novex XCell II Surelock; IBI Scientific Universal Protein Systems

Thermo Scientific™ Precise Tris-Glycine Gels are long shelf-life, fast run-time polyacrylamide gels for SDS-PAGE that use the popular Laemmli buffer system and commercial gel tanks to provide outstanding protein resolution and separation.

Precise Tris-Glycine Gels are cast in durable 10 × 8cm plastic cassettes to fit in Mini-PROTEAN™ gel tanks. The gels are individually packaged in easy-to-open plastic pouches and are ready for electrophoresis with no comb or tape to remove. Following electrophoresis, the easy-open cassette design allows gels to be removed damage-free without special tools. Precise Gels in the 10 × 8cm size are available in gradient (4 to 20% and 8 to 16%) and fixed (8%, 10% and 12%) concentrations and in 10-well, 12-well and 15-well formats.

Highlights:

  • High-performance – Tris-glycine running buffer produces excellent separation and high-resolution protein bands

  • Long shelf-life – up to 12-month stability ensures consistent performance for the duration of each package of gels

  • Fast – 45-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes; stains quickly with coomassie and silver stains

  • Compatible – these 10 x 8cm cassettes fit Bio-Rad™ Mini-PROTEAN™ and Hoefer™ Mighty Small II gel tanks, as well as other similar tanks

  • Easy to load – sample wells reinforced with plastic to eliminate damage when loading and well-to-well contamination

Specifications:

  • Cassette Dimensions: 10cm x 8.0cm x 5mm

  • Gel Dimensions: 8cm x 6.8cm x 1mm

  • Storage Conditions: 4°C

  • Shelf Life: 12 months from date of purchase

  • Stacking Gel: 4%

  • Buffer System in Gel: Tris-HCl, pH 7

  • SDS: None

  • Required Running Buffer: Tris-glycine-SDS

  • Recommended Sample Buffer: Tris-HCl-SDS or LDS-SDS

  • Gel Tank Compatibility (10cm x 8cm x 5mm): Bio-Rad Mini-PROTEAN II and 3; Bio-Rad Mini-PROTEAN Tetra Cell; Hoefer Tall Mighty Small (SE 280); Hoefer Mighty Small II (SE 260); IBI Scientific Universal Protein Systems; For gels that are compatible with other tanks, see our 10 x 10cm Gels

TRUSTED_SUSTAINABILITY

Specifications

Description 8% Tris-Glycine Gels, 10 x 8cm, 12-well
Gel Percentage 8%
Gel Thickness 0.5 cm
Gel Type Tris-Glycine
Separation Range 45 to 205 kDa
Separation Type Denaturing, Native
Wells 12-well
Formulation Precast 8% polyacrylamide gels for SDS-PAGE
Quantity 10/Pk.
Sufficient For 12 samples per gel, 30uL capacity per well
Well Design Standard
Length (Metric) 10 cm
Width (Metric) 8 cm
Product Type Homogeneous Page
For Use With (Equipment) Novex XCell I and II, Novex XCell II Surelock, IBI Scientific Universal Protein Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.