missing translation for 'onlineSavingsMsg'
Learn More

Alla France™ Precision Alcoholmeter

Product Code. 15824034
Change view
Click to view available options
Length (Metric):
290 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15824034 290 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15824034 Supplier Alla France™ Supplier No. 0530FC050/20QP
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Alla France™ Precision Alcoholmeter is used to determine the amount of alcohol in a liquid.

  • Calibration temperature 20°C
  • High precision
  • For use in the distillery and wine industry

Specifications

Type Range Finder Alcoholometer
Accuracy ±0.5
Increments 0.5
Length (Metric) 290 mm
Description 0 to 50 % Vol.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.