missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Precision™ Circulating Water Baths

Product Code. 15355867 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
19 L
35 L
89 L
Model:
Precision CIR 19
Precision CIR 35
Precision CIR 89
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Model Unit Size
15355867 89 L Precision CIR 89 Each
15335867 19 L Precision CIR 19 Each
15183804 19 L Precision CIR 19 Each
15345867 35 L Precision CIR 35 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15355867 Supplier Thermo Scientific™ Supplier No. TSCIR89
  • ONLINE EXCLUSIVE PRICE
    3930.00 €
    3740.00 € / Pack of 1
    Save 190.00 €
    4% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on these circulating water baths. Featuring advanced temperature controller.

Ensure temperature uniformity and control with theseThermo Scientific™ Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.

  • Achieve enhanced temperature uniformity with perimeter-directed water flow
  • Easily clean and maintain bath with coil-free internal design
  • Optimize scheduling with auto-on and auto-off timers
  • Accommodate taller labware with new hinged lid and extended height
  • Help prevent bath overheating and damage with low-fluid protection
  • Easily operate and monitor with icon-based graphical display
  • Protect your work with audible alarms
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 23.5 gal.
Controller Type Digital
Capacity (Metric) 89 L
Certifications/Compliance UL, CE
Communications Digital
Temperature Range Ambient +5° to 100°C
Display Monochrome LCD
Heating Capacity 1500 w
Includes 1 Stainless Steel Gable cover and Rubber Duck
Material 304 Stainless Steel
Model Precision CIR 89
Temperature Presets 4
Temperature Stability ±0.1°C
Temperature Uniformity ±0.05°C
Dimensions (D x W x H) Interior 19 x 36 x 9.5 in.(483 x 914 x 241 mm)
Dimensions (L x W x H) 21.5 x 45.7 x 11.8 in. (546 x 1160 x 300 mm)
Electrical Requirements Global Voltage 100/115 V 200/230 V 50/60 Hz
Amperage 11.2-13.1 / 5.4-6.3 (120V/230V)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.