missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ High Precision, Digital Dry Block Heating System

Product Code. p-7228784 Shop All Fisherbrand Products
Click to view available options
No. of Blocks:
1 Block
2 Blocks
4 Blocks
Height (Metric):
100 mm
120 mm
Wattage:
150 W
300 W
600 W
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17275546

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides great temperature control and a wide choice of interchangeable blocks for maximum flexibility. These Dry Block Heating Systems combine ideal temperature control and uniformity over a temperature range of ambient +5°C to 130°C.

  • High power heater for fast heat up - 25°C to 100°C in only 15 minutes
  • Simple-to-use rotor plus two keys provide access to the interactive interface for fast, accurate set-up and use
  • Clear digital display is easy to read from a distance for instant reassurance
  • Convenient timer facility, with audible buzzer, for reaction timing and function timing, e.g. delayed heater switch-on/turn-off
  • Wide range of interchangeable blocks enables you to mix and match sample tubes as desired with extraction tool supplied as standard for easy and safe removal of blocks
  • Optional safety cover protects samples from contamination and users from accidental contact with hot blocks
  • Overtemperature cut-out protects your samples and your workplace
  • High quality, robust construction in streamlined coolwall aluminum and chemical-resistant plastic is durable in demanding environments
  • Compact footprint to optimize use of bench space
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 1.7 to 16.9 oz.
Autoclavable Not Autoclavable
Capacity (Metric) 50 to 500 mL
Chamber Material Aluminum
For Use With (Equipment) Interchangeable Blocks
Material Chemical Resistant Plastic
No. of Places 1
Plug Type EU, UK
Relative Humidity 80%
Temperature Range (Metric) 5°C to 130°C
Type Dry Bath Block
For Use With (Application) Enzyme Digestion and Activity Studies, Nucleic Acid Hybridization
Heat Up Time 20 min. (25°C to 100°C)
No. of Blocks 1 Block
Temperature Stability 0.1°C at 37°C
Temperature Uniformity 0.1°C at 37°C
Timer 1 to 999 min.
Warranty 3 Years
Width (English) 7.9 in.
Width (Metric) 200 mm
Height (English) 4.7 in.
Height (Metric) 120 mm
Depth (English) 4.3 in.
Depth (Metric) 230 mm
Voltage 220/240 V
Wattage 150 W
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.