Learn More
MP BIOMEDICALS Precision Tissue Wipers
Product Code. 30928576 Shop All MP Biomedicals Products
Change view
Click to view available options
Product Type:
Low-lint Wiper
Unit Size:
Pack of 280
Product Code. Product Type Unit Size
30928576 Low-lint Wiper Pack of 280
1 options
Product Code. 30928576 Supplier MP BIOMEDICALS Supplier No. 07DP1030
  • ONLINE EXCLUSIVE PRICE
    18.85 €
    17.85 € / Pack of 280
    Save 1.00 €
    5% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Low-lint wipers for delicate applications.
  • Kim Wipes designed for delicate laboratory and manufacturing tasks requiring minimal lint and particle generation.
  • Ideal for cleaning sensitive surfaces, instruments, and optical equipment without causing damage.
  • Supplied for research or further manufacturing use only; not intended for direct human use.
  • Durable and absorbent, providing efficient wiping and contamination control in precise laboratory environments.
Specifications
Product Type Low-lint Wiper
Material Tissue