missing translation for 'onlineSavingsMsg'
Learn More

Prednisolone 21-Hemisuccinate Sodium Salt, TRC

Product Code. 30428471
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428471 1 g -
30421283 5 g -
30412596 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428471 Supplier Toronto Research Chemicals Supplier No. TRCP7039131G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1715-33-9
Molecular Formula C25H31NaO8
Molecular Weight (g/mol) 482.5
Synonym Di-Adreson F-aquosum, Meticortelane, Meticotelone Soluble, NSC 9151, Prednine, Prednisolone 21-hemisuccinate Sodium, Prednisolone 21-hemisuccinate Sodium Salt, Prednisolone 21-Sodium succinate, Prednisolone 21-Succinate Sodium, Prednisolone 21-Succinate Sodium Salt, Prednisolone Hemisuccinate Sodium Salt, Prednisolone Sodium 21-Hemisuccinate, Prednisolone Sodium Hemisuccinate, Prednisolone Sodium Succinate, Predonine Soluble, Sodium prednisolone 21-hemisuccinate, Solu-Decortin, Solu-Decortin H, Solu-Delta-Cortef, 21-Monoester with 11β,17,21-Trihydroxypregna-1,4-diene-3,20-dione Succinic Acid Monosodium Salt, 11β,17,21-Trihydroxy-pregna-1,4-diene-3,20-dione 21-Succinate Monosodium Salt
IUPAC Name sodium;4-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoate
SMILES [Na+].C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)COC(=O)CCC(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C25H32O8.Na/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30;/h7,9,11,16-18,22,27,32H,3-6,8,10,12-13H2,1-2H3,(H,29,30);/q;+1/p-1/t16-,17-,18-,22+,23-,24-,25-;/m0./s1
Formula Weight 482.1917
Chemical Name or Material Prednisolone 21-Hemisuccinate Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.