missing translation for 'onlineSavingsMsg'
Learn More

Prednisolone, 99%, Thermo Scientific Chemicals

Product Code. 13521640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
Product Code. Quantity Packaging unitSize
13521640 25 g Glass Bottle 25g
13501640 1 g Glass Bottle 1g
13511640 5 g Glass Bottle 5g
3 options
This item is not returnable. View return policy

Product Code. 13521640

Brand: Thermo Scientific Acros 449470250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prednisolone, or metacortandralone, CAS # 50-24-8, is a synthetic glucocorticoid.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Prednisolone is a derivative of prednisone with an oxo to beta-hydroxy reduction at position 11

• Prednisolone binds cell surface receptors and is transported to the nucleus, where it binds certain nuclear receptors responsible for transcription of proinflammatory cytokines

Applications

• Prednisolone can be used for laboratory research on tumor growth and prevention

• Prednisolone may also be used for developmental and endocrine research, in vivo or in vitro

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-24-8
Molecular Formula C21H28O5
Molecular Weight (g/mol) 360.44
InChI Key OIGNJSKKLXVSLS-VWUMJDOOSA-N
Synonym prednisolone, metacortandralone, hydroretrocortine, predonine, deltacortril, meticortelone, deltahydrocortisone, prenolone, hydroretrocortin, delta-cortef
PubChem CID 5755
ChEBI CHEBI:8378
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
SMILES CC12CC(C3C(C1CCC2(C(=O)CO)O)CCC4=CC(=O)C=CC34C)O

Specifications

Melting Point 240.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 25 g
Merck Index 15,7841
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol, dioxane, slightly soluble in acetone, chloroform,, ethylacetate
Formula Weight 360.44
Percent Purity 99%
Chemical Name or Material Prednisolone
compliance-icons
Product Identifier
  • Prednisolone
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.