missing translation for 'onlineSavingsMsg'
Learn More

Prednisolone, MedChemExpress

Product Code. 16479837
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479837 5 g 5g
16471679 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479837 Supplier Medchem Express Supplier No. HY17463
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Prednisolone is a potent, orally active corticosteroid and a glucocorticoid. Prednisolone possesses about four times the anti-inflammatory activity of hydrocortisone while causing less salt and water retention. Prednisolone can be used for ocular, anti-inflammatory research.

Prednisolone is a potent, orally active corticosteroid and a glucocorticoid. Prednisolone possesses about four times the anti-inflammatory activity of hydrocortisone while causing less salt and water retention. Prednisolone can be used for ocular, anti-inflammatory research.

Specifications

Chemical Name or Material Prednisolone
CAS 50-24-8
Color White
Purity Grade Notes Research
Molecular Formula C21H28O5
Quantity 5 g
Solubility Information DMSO : 62.5 mg/mL (173.40 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.28 mM; Need ultrasonic)
SMILES C[C@@]1([C@@]2(O)C(CO)=O)[C@](CC2)([H])[C@@](CCC3=CC4=O)([H])[C@]([C@]3(C=C4)C)([H])[C@@H](O)C1
Molecular Weight (g/mol) 360.44
Formula Weight 360.44
Percent Purity 99.77%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.