missing translation for 'onlineSavingsMsg'
Learn More

Prednisolone, TRC

Product Code. 30006357
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006357 10 g 10g
30010989 50 g 50g
18072130 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006357 Supplier Toronto Research Chemicals Supplier No. TRCP70374010G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-24-8
Molecular Formula C21 H28 O5
Molecular Weight (g/mol) 360.44
Synonym Prednisolone Acetate Imp. B (EP), Hydrocortisone Imp. A (EP), Pregna-1,4-diene-3,20-dione, 11,17,21-trihydroxy-, (11β)-, Pregna-1,4-diene-3,20-dione, 11β,17,21-trihydroxy- (8CI), (11β)-11,17,21-Trihydroxypregna-1,4-diene-3,20-dione, 1,2-Dehydrohydrocortisone, 1,4-Pregnadiene-11β,17α,21-triol-3,20-dione, 1,4-Pregnadiene-3,20-dione-11β,17α,21-triol, 1-Dehydrohydrocortisone, 11β,17,21-Trihydroxypregna-1,4-diene-3,20-dione, 11β,17α,21-Trihydroxypregna-1,4-diene-3,20-dione, Aprednislon, Co-Hydeltra, Codelcortone, Cortalone, Decaprednil, Decortin H, Delcortol, Delta F, Delta-Cortef, Delta-Ef-Cortelan, Delta-stab, Deltacortenol, Deltacortril, Deltacortril Enteric, Deltahydrocortisone, Deltasolone, Deltisilone, Di-Adreson F, Dicortol, Donisolone, Eazolin D, Fernisolone, Flamasone, Hostacortin H, Hydeltra, Hydeltrone, Hydrodeltalone, Hydrodeltisone, Hydroretrocortin, Hydroretrocortine, Klismacort, Metacortandralone, Meti-Derm, Meticortelone, Millipred, NSC 9120, NSC 9900, Panafcortelone, Paracortol, Precortalon, Precortancyl, Precortilon, Precortisyl, Predne-Dome, Prednelan, Predni-Dome, Prednicare, Prednicen, Predniretard, Prednisolona Syntex, Prednisolone, Predonin, Predonine, Preflam, Prelone, Prenolone, Scherisolon, Solone, Steran, Sterane, Sterolone, Supercortisol, Ulacort, Ultracorten H, Ultracortene H, Vetsolone, δ1-Cortisol, δ1-Dehydrocortisol, δ1-Dehydrohydrocortisone, δ1-Hydrocortisone
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

Quantity 10 g
InChI Formula InChI=1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1
Formula Weight 360.1937
Percent Purity >95%
Chemical Name or Material Prednisolone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.