missing translation for 'onlineSavingsMsg'
Learn More

Shermond™ Premier Latex Examination Gloves

Product Code. 12818210
Encompass_Preferred
Click to view available options
:
Size: Large
This item is not returnable. View return policy

Product Code. 12818210

Brand: Shermond™ 6603L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Shermond™ Premier Latex Examination Gloves are powder free and have an enhanced textured surface to provide excellent touch sensitivity and grip, ideal for working with fluids, instruments or wet glassware.

  • For skin protection when handling wet or dry abrasive materials and to protect against non-hazardous liquids
  • Powder free
  • Enhanced textured surface
  • Excellent touch sensitivity and grip
  • Ideal for working with fluids, instruments or wet glassware

Specifications

Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Hand Ambidextrous
Latex Free No
Material Latex
Powdered Powder Free
Product Type Examination Gloves
Size Large
Sterility Non-sterile
Thickness (Metric) 0.15 mm
For Use With (Application) Examination duties
Resistant To Non-hazardous liquids
Length (Metric) 240 mm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.