Learn More
Fisherbrand™ Premium-Line Low Retention Pipette Tips
Product Code. 10283882 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 20 μL
1 to 200 μL
100 to 1000 μL
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 960
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Format unitSize
10283882 1 to 200 μL Non-sterile Bulk Pack of 1000
10050892 0.1 to 10 μL Non-sterile Bulk Pack of 1000
10365862 0.5 to 20 μL Non-sterile Bulk Pack of 1000
10758294 0.1 to 10 μL Sterile Rack Pack of 960
10325912 0.5 to 20 μL Non-sterile Rack Pack of 960
10521973 1 to 200 μL Sterile Rack Pack of 960
10671973 0.5 to 20 μL Sterile Rack Pack of 960
10778484 1 to 200 μL Non-sterile Rack Pack of 960
10571963 100 to 1000 μL Non-sterile Rack Pack of 960
10204462 100 to 1000 μL Sterile Rack Pack of 960
10738104 0.1 to 10 μL Non-sterile Rack Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10283882
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Combines the advantages of standard tips with an innovative low retention surface to ensure maximum sample recovery, minimal sample loss, and high-precision pipetting. Fisherbrand™ Premium-Line Low Retention Pipette Tips also prevent sample contamination with conical tip opening.
  • Optimized cone geometry provides a secure fit
  • Graduation marks on tip at standard volumes
  • Precise pipetting without loss of sample
  • Conical tip opening prevents drop formation at the end of the tip
  • Elimination of sample contamination
  • Made of autoclavable, non-wettable, virgin polypropylene
  • Tips are autoclavable fo sterile handling
Specifications
Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (Metric) 50.8mm
Certifications/Compliance CE, IVD
Sterilization Method Autoclave
Format Bulk
Graduated Yes
Material Virgin Polypropylene
Graduations Standard Volumes