missing translation for 'onlineSavingsMsg'
Learn More

Prenyl Acetate, TRC

Product Code. 30416927
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416927 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416927 Supplier Toronto Research Chemicals Supplier No. TRCP71279050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prenyl Acetate
CAS 1191-16-8
Molecular Formula C7 H12 O2
Quantity 50 g
Synonym 1-Acetoxy-3-methyl-2-butene, 2-Methyl-4-acetyloxy-2-butene, 3,3-Dimethylallyl Acetate, 3-Methyl-1-acetoxy-2-butene, 3-Methyl-2-buten-1-ol Acetate, 3-Methyl-2-buten-1-yl Acetate, 3-Methyl-2-butenyl Acetate, Acetic Acid 3-Methyl-2-butenyl Ester, Dimethylallyl Acetate, Isopent-2-enyl Acetate, γ,γ-Dimethylallyl Acetate
InChI Formula InChI=1S/C7H12O2/c1-6(2)4-5-9-7(3)8/h4H,5H2,1-3H3
SMILES CC(=CCOC(=O)C)C
IUPAC Name 3-methylbut-2-enyl acetate
Molecular Weight (g/mol) 128.169
Formula Weight 128.084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.