missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ Elution Buffer

Product Code. 30000688 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000688 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000688 Supplier Applied Biosystems™ Supplier No. 4400784C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PrepSEQ Elution Buffer, a component of the PrepSEQ Nucleic Acid Extraction Kit, is offered separately here for those kit users who need additional elution buffer.

PrepSEQ Elution Buffer, a component of the PrepSEQ Nucleic Acid Extraction Kit (Cat. No. 4413686), is offered separately here for those kit users who need additional elution buffer. As with other products in the Pharma Analytics Sample Preparation product line, PrepSEQ Elution Buffer:

  • Is completely supported by our global manufacturing and distribution centers
  • Has a Certificate of Analysis available
  • Has a shelf-life of up to 12 months
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Elution Buffer
Physical Form Liquid
Quantity 25 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.