missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ Residual DNA Sample Preparation Kit

Product Code. 30000671 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000671 100 preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000671 Supplier Applied Biosystems™ Supplier No. 4413686
  • ONLINE EXCLUSIVE PRICE
    1270.00 €
    1108.00 € / Pack of 1
    Save 162.00 €
    12% Off
View all promotions
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems PrepSEQ Residual DNA Sample Preparation Kit is optimized for highly efficient DNA recovery from complex mixtures of proteins, buffers, and salts which are typical of samples that need to be quantitated for residual host cell line DNA.

The Applied Biosystems PrepSEQ Residual DNA Sample Preparation Kit is optimized for highly efficient DNA recovery from complex mixtures of proteins, buffers, and salts which are typical of samples that need to be quantitated for residual host cell line DNA. Quantitative and consistent recovery of DNA can be obtained from challenging matrices, including samples that have high protein concentration, low pH, or high salt.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Includes buffers, magnetic beads and enzymes for DNA extraction for residual DNA quantitation for 100 reactions
Isolation Technology Magnetic Bead
Scale Mini
Sample Type Biopharmaceutical Production Samples
Final Product Type Genomic DNA
For Use With (Application) Impurity Testing
High-throughput Compatibility Not High-throughput Compatible (Manual), Automated Protocols
No. of Reactions 100 preps
Product Line PrepSEQ
Quantity 100 preps
Sample Volume 100 μL
Target Genomic DNA
Test Time 2 hr.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • PrepSEQ Residual DNA Sample Preparation Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: N-LAUROYLSARCOSINE SODIUM SALT, Thiocyanic acid, Proteinase,Glycerin

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.