missing translation for 'onlineSavingsMsg'
Learn More

Presept™ Disinfectant Tablets 2.5g

Product Code. 12368667
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
2.5 g
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12368667 2.5 g Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12368667 Supplier Johnson Johnson Medical Supplier No. SPR25
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rapid-acting disinfectant tablets.

The solution is rapid acting, and is effective against bacteria, spores, fungi, algae, protozoa and viruses, (including AIDS virus) and is resistant to inactivation by organic soilage.
These 2.5g tablets contain sodium dichloroisocyanurate (NaDCC) in an effervescent base. The tablets dissolve in water to provide disinfectant solutions of predetermined concentrations, without the need to weigh or measure the active ingredient. The solution is compatible with anionic and non-ionic detergents. One tablet dissolved in 10L of water provides 140ppm available chlorine. Dilution instructions are provided in each pack. Supplied in resealable plastic drum containing 100 tablets.

Use biocides safely. Always read the label and product information before use.

Specifications

Product Type Disinfectant Tablet
Format Tablet
Quantity 100/Pk.
Volume (Metric) 2.5 g
compliance-icons compliance-icons
Product Identifier
  • PRESEPT Disinfectant Tablets & Granules
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contain: sodium chloride, sodium carbonate, adipic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.