Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Product Code. 11367151 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Filter
Filter with Stabilizer
Prefilter
Prefilter MQ Cartridge
Pretreatment Cartridge
Pretreatment Filter
Stabilizing Cartridge
Unit Size:
1 unit
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11367151 Pretreatment Filter Each
11327151 Pretreatment Cartridge Each
11766259 Prefilter MQ Cartridge Each
15322178 Prefilter 1 unit
12756208 Filter with Stabilizer Each
11387101 Stabilizing Cartridge Each
12818111 Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11367151 Supplier Thermo Scientific™ Supplier No. 09.4003
Add to basket
Add to basket
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Smart2Pure, LabTower RO, LabTower TII
Type Pretreatment Filter
Includes 1 μm Filter, Filter Housing