missing translation for 'onlineSavingsMsg'
Learn More

Primaquine diphosphate, 98%

Product Code. 15678685 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15678685 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15678685 Supplier Thermo Scientific Acros Supplier No. 170870050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63-45-6 | C15H21N3O·2H3PO4 | 455.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 63-45-6
Melting Point 205.0°C to 206.0°C
Infrared Spectrum Authentic
Molecular Formula C15H21N3O·2H3PO4
Quantity 5 g
Merck Index 15,7863
Synonym primaquine phosphate, n4-6-methoxyquinolin-8-yl pentane-1,4-diamine phosphate, n-5-aminopentan-2-yl-6-methoxyquinolin-8-amine; phosphoric acid, 4-n-6-methoxyquinolin-8-yl pentane-1,4-diamine; phosphoric acid, primaquine diphosphate, chembl994, phosphoric acid; primaquine, 1, n4-6-methoxy-8-quinolinyl-, phosphate 1:2, n-5-aminopentan-2-yl-6-methoxyquinolin-8-amine phosphoric acid, n4-6-methoxyquinolin-8-yl pentane-1,4-diamine; phosphoric acid
Solubility Information Solubility in water: moderately soluble.
InChI Key GJOHLWZHWQUKAU-UHFFFAOYSA-N
SMILES CC(CCCN)NC1=C2C(=CC(=C1)OC)C=CC=N2.OP(=O)(O)O.OP(=O)(O)O
IUPAC Name 4-N-(6-methoxyquinolin-8-yl)pentane-1,4-diamine;phosphoric acid
Molecular Weight (g/mol) 455.34
PubChem CID 6135
Formula Weight 455.34
Percent Purity 98%
Chemical Name or Material Primaquine diphosphate
Show More Show Less
compliance-icons
Product Identifier
  • Primaquine diphosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: Primaquine bis(phosphate)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.