missing translation for 'onlineSavingsMsg'
Learn More

Primaquine Diphosphate, TRC

Product Code. 30137968
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137968 1 g 1g
30011647 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137968 Supplier Toronto Research Chemicals Supplier No. TRCP7335001G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-45-6
Molecular Formula C15 H21 N3 O . 2 H3 O4 P
Molecular Weight (g/mol) 455.34
Synonym Primaquine diphosphate, (4RS)-N4-(6-Methoxyquinolin-8-yl)pentane-1,4-diamine bisphosphate, Quinoline, 8-[(4-amino-1-methylbutyl)amino]-6-methoxy-, diphosphate, Quinoline, 8-[(4-amino-1-methylbutyl)amino]-6-methoxy-, phosphate (1:2), 8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline di(dihydrogen phosphate), 8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline diphosphate, Camden, NSC 149765, Pharmaniaga, Primaquine phosphate, WR 2975
IUPAC Name 4-N-(6-methoxyquinolin-8-yl)pentane-1,4-diamine;phosphoric acid
SMILES COc1cc(NC(C)CCCN)c2ncccc2c1.OP(=O)(O)O.OP(=O)(O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H21N3O.2H3O4P/c1-11(5-3-7-16)18-14-10-13(19-2)9-12-6-4-8-17-15(12)14;2*1-5(2,3)4/h4,6,8-11,18H,3,5,7,16H2,1-2H3;2*(H3,1,2,3,4)
Formula Weight 455.1223
Chemical Name or Material Primaquine diphosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.