missing translation for 'onlineSavingsMsg'
Learn More

Primary Color Solution, Blue, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11865853 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11865853 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11865853 Supplier Reagecon Supplier No. EPBS01
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Used to calibrate, control, qualify and validate color measurement instruments. Primary Color Solution, Blue, EP Grade, Reagecon™ is a ready-to-use colorimetric solution presented in high-quality, tamper-evident bottles. It is prepared with copper sulfate in accordance with Eu. Ph. Chapter 2.

This Primary Color Solution is a ready-to-use coloration standard presented in high-quality, tamper-evident bottles for use as a calibration and/or quality control standard. It is prepared with copper sulfate, hydrochloric acid and water.
  • Manufactured according to European Pharmocopoeia Chapter 2 guidelines with a Balance Certificate of Calibration issued in accordance with ISO 17025
  • Prepared gravimetrically on a weight/weight basis using OIML traceable weights
  • Concentration verified using a high-performance spectrophotometer calibrated using ISO Guide 34 accredited spectrophotometric standards
  • The Primary Color Solutions can be combined in precise ratios to form the Standard Color Solutions

Specifications

CAS 7758-98-7
Molecular Formula CuO4S
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)[O-].[Cu+2]
IUPAC Name copper;sulfate
Molecular Weight (g/mol) 159.602
PubChem CID 24462
ChEBI CHEBI:23414
Formula Weight 162.20g/mol
Color Blue
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Copper(II) Sulfate
Show More Show Less
compliance-icons
Product Identifier
  • PRIMARY BLUE SOLUTION
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: copper sulphate, hydrochloric acid 37%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.