missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Primate Heart Membrane Tissue Lysate (Adult Membrane Normal)

Product Code. 18281910
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18281910 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18281910 Supplier Novus Biologicals™ Supplier No. NBL126476
  • ONLINE EXCLUSIVE PRICE
    527.00 €
    497.70 € / 0.1mg
    Save 29.30 €
    5% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Primate Heart Membrane Tissue Lysate (Adult Membrane Normal) is derived from primate.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot Electrophoresis
Tissue Heart
Quantity 0.1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Membrane Protein - Monkey (Rhesus) Normal Tissue: Heart
Cellular Fraction Membrane
Tissue Condition Normal
Cross Reactivity Monkey
Host Species Primate
Product Type Primate Heart Membrane Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.