missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRIMER 200 NMOL

Product Code. 10513633
Change view
Click to view available options
200 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10513633 200 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 10513633 Supplier Invitrogen™ Supplier No. 10629020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Custom Primer

Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Normal or Reversed-phase Chromatography, HPLC or PAGE
Description Custom Primer, 20 nmole, Purified by Normal or Reversed-phase Chromatography, HPLC or PAGE, Used to Design a Custom DNA Oligonucleotide, With any of a Variety of 3' and 5' Modification
Product Type Custom Primer
Quantity 200 nmol
Grade HPLC
For Use With (Application) Design a Custom DNA Oligonucleotide, with any of a variety of 3' and 5' Modification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.